2-(Trifluoromethyl)phenylacetonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(Trifluoromethyl)benzyl cyanide1-(Cyanmethyl)-2-(trifluormethyl)benzene

PC7709-1G
76627-500EA 13.39 USD
76627-500
2-(Trifluoromethyl)phenylacetonitrile 97%
2-(Trifluoromethyl)phenylacetonitrile
Formula: C₉H₆F₃N
MW: 185.15 g/mol
Boiling Pt: 92 °C/11mmHg
Melting Pt: 32…35 °C
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00001903
CAS Number: 3038-47-9
EINECS: 221-239-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR