3,5-Difluoro-4-hydroxybenzaldehyde 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC8951-25G PC8951-1G PC8951-5G
76609-736EA 143 USD
76609-736 76609-732 76609-734
3,5-Difluoro-4-hydroxybenzaldehyde 99%
3,5-Difluoro-4-hydroxybenzaldehyde
Formula: C₇H₄F₂O₂
Melting Pt: 119…125 °C
Storage Temperature: Ambient
MDL Number: MFCD06797917
CAS Number: 118276-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR