3'-Chloro-2',6'-difluoroacetophenone 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(3-Chloro-2,6-difluorophenyl)ethanone

PC9122-25G PC9122-1G PC9122-5G
76610-126EA 391 USD
76610-126 76610-124 76610-128
3'-Chloro-2',6'-difluoroacetophenone 98%
3'-Chloro-2',6'-difluoroacetophenone
Formula: C₈H₅ClF₂O
MW: 190.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631329
CAS Number: 177942-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR