4-Chloro-2,6-difluorobenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC53003-1G PC53003-5G
76626-986EA 90 USD
76626-986 76626-984
4-Chloro-2,6-difluorobenzonitrile 98%
4-Chloro-2,6-difluorobenzonitrile
Formula: C₇H₂ClF₂N
MW: 173.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660237
CAS Number: 886500-41-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR