3-Bromo-2-fluorobenzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC6975-25G PC6975-1G PC6975-5G
76608-016EA 13.39 USD
76608-016 76608-020 76608-018
3-Bromo-2-fluorobenzonitrile
3-Bromo-2-fluorobenzonitrile
Formula: C₇H₃BrFN
MW: 200.01 g/mol
Melting Pt: 37…39 °C
Storage Temperature: Ambient
MDL Number: MFCD06657980
CAS Number: 840481-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR