3-(Trifluoromethyl)benzophenone 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC7569-5G PC7569-25G
76608-654EA 28 USD
76608-654 76608-656
3-(Trifluoromethyl)benzophenone 98%
3-(Trifluoromethyl)benzophenone
Formula: C₁₄H₉F₃O
MW: 250.22 g/mol
Boiling Pt: 121…123 °C/1.5mmHg
Melting Pt: 51…52 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000389
CAS Number: 728-81-4
EINECS: 211-973-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR