α,α,α-Trifluoro-m-toluic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-(Trifluoromethyl)benzoic acid

PC7530-500G PC7530-1KG PC7530-100G
76608-620EA 170.98 USD
76608-620 76608-618 76608-622
α,α,α-Trifluoro-m-toluic acid 98%
α,α,α-Trifluoro-m-toluic acid
Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 237…240 °C
Melting Pt: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00002519
CAS Number: 454-92-2
EINECS: 207-230-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR