2-Fluoro-m-xylene 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2-Fluoro-1,3-dimethylbenzene1-Fluoro-2,6-dimethylbenzene

PC4381-5G PC4381-25G PC4381-100G
76613-972EA 54 USD
76613-972 76613-974 76613-970
2-Fluoro-m-xylene 98%
2-Fluoro-m-xylene
Formula: C₈H₉F
MW: 124.16 g/mol
Boiling Pt: 147…148 °C
Density: 0.988 g/cm³ (25 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00039215
CAS Number: 443-88-9
EINECS: 207-144-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR