α'-Bromo-α,α,α-trifluoro-m-xylene 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-(Trifluoromethyl)benzyl bromide1-(Bromomethyl)-3-(trifluoromethyl)benzene3-(Bromomethyl)benzotrifluoride

PC7632-250G PC7632-25G PC7632-100G
76627-476EA 27 USD
76627-476 76627-474 76627-472
α'-Bromo-α,α,α-trifluoro-m-xylene 97%
α'-Bromo-α,α,α-trifluoro-m-xylene
Formula: C₈H₆BrF₃
MW: 239.03 g/mol
Boiling Pt: 69 °C/4mmHg
Density: 1.565 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00000395
CAS Number: 402-23-3
EINECS: 206-938-9
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR