α,α,α,4-Tetrafluorotoluene 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC3600-500G PC3600-25G PC3600-100G
76613-800EA 13 USD
76613-800 76613-802 76613-798
α,α,α,4-Tetrafluorotoluene 99%
α,α,α,4-Tetrafluorotoluene
Formula: C₇H₄F₄
MW: 164.1 g/mol
Boiling Pt: 102…105 °C
Melting Pt: –42 to –41 °C
Density: 1.293 g/cm³ (25 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00000399
CAS Number: 402-44-8
EINECS: 206-944-1
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR