3-Fluorotoluene 99%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 1-Fluoro-3-methylbenzene

PC4320-1KG PC4320-500G PC4320-100G
76613-922EA 64 USD
76613-922 76613-924 76613-920
3-Fluorotoluene 99%
3-Fluorotoluene

Intermediate for the production of Ciprofloxacin.

Formula: C₇H₇F
Boiling Pt: 114…116 °C
Melting Pt: –87 °C
Density: 0.991 g/cm³ (25 °C)
Flash Pt: 9 °C
Storage Temperature: Ambient
MDL Number: MFCD00000339
CAS Number: 352-70-5
EINECS: 206-524-8
UN: 2388
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR