α,α,α,3-Tetrafluorotoluene 99%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 3-Fluorobenzotrifluoride1-Fluoro-3-(trifluoromethyl)benzene

PC3590-100G PC3590-1KG PC3590-250G
76613-796EA 37 USD
76613-796 76613-792 76613-794
α,α,α,3-Tetrafluorotoluene 99%
α,α,α,3-Tetrafluorotoluene
Formula: C₇H₄F₄
MW: 164.1 g/mol
Boiling Pt: 101…102 °C
Density: 1.302 g/cm³ (25 °C)
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00000382
CAS Number: 401-80-9
EINECS: 206-933-1
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR