4-Bromo-2-fluorotoluene 95%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 1-Bromo-3-fluoro-4-methylbenzene

PC56731-1KG PC56731-100G PC56731-500G
76602-570EA 27 USD
76602-570 76602-574 76602-572
4-Bromo-2-fluorotoluene 95%
4-Bromo-2-fluorotoluene
Formula: C₇H₆BrF
Storage Temperature: Ambient
MDL Number: MFCD00013551
CAS Number: 51436-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR