2-Chloro-6-(trifluoromethoxy)benzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC302444-1G
76597-754EA 36 USD
76597-754
2-Chloro-6-(trifluoromethoxy)benzonitrile
2-Chloro-6-(trifluoromethoxy)benzonitrile
Formula: C₈H₃ClF₃NO
MW: 221.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18393805
CAS Number: 1261779-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR