2-Fluoro-4-methoxybenzamide 97%

Supplier: APOLLO SCIENTIFIC

PC302803-5G PC302803-10G PC302803-1G
76598-710EA 44 USD
76598-710 76598-708 76598-712
2-Fluoro-4-methoxybenzamide 97%
2-Fluoro-4-methoxybenzamide
Formula: C₈H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD01117440
CAS Number: 874804-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR