3,5-Difluoro-2-ethoxybenzonitrile 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: 2-Ethoxy-3,5-difluorobenzonitrile

PC302679-1G PC302679-5G PC302679-10G
76626-086EA 150.76 USD
76626-086 76626-082 76626-084
3,5-Difluoro-2-ethoxybenzonitrile 98%
3,5-Difluoro-2-ethoxybenzonitrile
Formula: C₉H₇F₂NO
Storage Temperature: Ambient
MDL Number: MFCD09258716
CAS Number: 1017779-80-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR