4-Chloro-3-ethoxy-2-fluorobenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC

PC302984-1G PC302984-5G PC302984-10G
76599-134EA 284 USD
76599-134 76599-138 76599-136
4-Chloro-3-ethoxy-2-fluorobenzaldehyde 98%
4-Chloro-3-ethoxy-2-fluorobenzaldehyde
Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD19687180
CAS Number: 1323966-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR