4-Methoxy-3-(trifluoromethoxy)benzonitrile 97%

Supplier: APOLLO SCIENTIFIC

PC302344-1G
76597-480EA 114 USD
76597-480
4-Methoxy-3-(trifluoromethoxy)benzonitrile 97%
4-Methoxy-3-(trifluoromethoxy)benzonitrile
Formula: C₉H₆F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD18398458
CAS Number: 1261653-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR