3-Ethoxy-2,6-difluorobenzyl alcohol 97%

Supplier: APOLLO SCIENTIFIC

Synonyms: (3-Ethoxy-2,6-difluorophenyl)methanol

PC302839-1G PC302839-5G PC302839-10G
76598-782EA 138 USD
76598-782 76598-780 76598-778
3-Ethoxy-2,6-difluorobenzyl alcohol 97%
3-Ethoxy-2,6-difluorobenzyl alcohol
Formula: C₉H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11519370
CAS Number: 1092461-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR