3-Methoxy-2-(trifluoromethyl)benzyl alcohol 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC302404-1G PC302404-10G PC302404-5G
76597-644EA 508 USD
76597-644 76597-642 76597-640
3-Methoxy-2-(trifluoromethyl)benzyl alcohol 98%
3-Methoxy-2-(trifluoromethyl)benzyl alcohol
Formula: C₉H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD18398589
CAS Number: 1261750-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR