3-Methoxy-2-(trifluoromethyl)benzaldehyde 98%

Supplier: APOLLO SCIENTIFIC

PC302094-1G PC302094-10G PC302094-5G
76596-782EA 623 USD
76596-782 76596-784 76596-786
3-Methoxy-2-(trifluoromethyl)benzaldehyde 98%
3-Methoxy-2-(trifluoromethyl)benzaldehyde
Formula: C₉H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD13185677
CAS Number: 1214384-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR