2-Bromo-3,6-difluorobenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

PC303025-5G PC303025-1G PC303025-10G
76599-210EA 201.88 USD
76599-210 76599-214 76599-212
2-Bromo-3,6-difluorobenzaldehyde
2-Bromo-3,6-difluorobenzaldehyde
Formula: C₇H₃BrF₂O
MW: 221.00 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12922618
CAS Number: 934987-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR