5-Methyl-2-(trifluoromethyl)benzaldehyde 98%

Supplier: APOLLO SCIENTIFIC

PC302893-1G PC302893-5G
76598-886EA 42 USD
76598-886 76598-888
5-Methyl-2-(trifluoromethyl)benzaldehyde 98%
5-Methyl-2-(trifluoromethyl)benzaldehyde
Formula: C₉H₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD06660294
CAS Number: 886502-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR