4-Fluoro-3-methyl-5-(trifluoromethyl)benzylamine 98%

Supplier: APOLLO SCIENTIFIC

PC302413-1G PC302413-5G
76613-236EA 98 USD
76613-236 76613-238
4-Fluoro-3-methyl-5-(trifluoromethyl)benzylamine 98%
4-Fluoro-3-methyl-5-(trifluoromethyl)benzylamine
Formula: C₉H₉F₄N
Storage Temperature: Ambient
MDL Number: MFCD22201070
CAS Number: 1373921-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR