2'-Methoxy-5'-(trifluoromethyl)acetophenone 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(2-Methoxy-5-(trifluoromethyl)phenyl)ethanone1-(2-Methoxy-5-trifluoromethylphenyl)ethanone

PC302929-1G PC302929-5G PC302929-10G
76598-984EA 283.25 USD
76598-984 76598-986 76598-988
2'-Methoxy-5'-(trifluoromethyl)acetophenone 98%
2'-Methoxy-5'-(trifluoromethyl)acetophenone
Formula: C₁₀H₉F₃O₂
MW: 218.17 g/mol
Melting Pt: 45…47 °C
Storage Temperature: Ambient
MDL Number: MFCD09832300
CAS Number: 503464-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR