3'-Fluoro-2'-(trifluoromethyl)acetophenone 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: 1-(3-Fluoro-2-(trifluoromethyl)phenyl)ethanone

PC302298-1G PC302298-5G
76597-392EA 1003.22 USD
76597-392 76597-394 76597-396
3'-Fluoro-2'-(trifluoromethyl)acetophenone 98%
3'-Fluoro-2'-(trifluoromethyl)acetophenone
Formula: C₉H₆F₄O
MW: 206.137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832249
CAS Number: 1017777-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR