2,6-Dichloro-3-(trifluoromethyl)benzamide 98%

Supplier: APOLLO SCIENTIFIC

PC302790-5G PC302790-10G PC302790-1G
76598-688EA 313 USD
76598-688 76598-690 76598-692
2,6-Dichloro-3-(trifluoromethyl)benzamide 98%
2,6-Dichloro-3-(trifluoromethyl)benzamide
Formula: C₈H₄Cl₂F₃NO
Melting Pt: 125…128 °C
Storage Temperature: Ambient
MDL Number: MFCD11519346
CAS Number: 53012-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR