2-Methoxy-6-(trifluoromethyl)benzylamine 98%

Supplier: APOLLO SCIENTIFIC

PC302260-1G
76613-200EA 328 USD
76613-200
2-Methoxy-6-(trifluoromethyl)benzylamine 98%
2-Methoxy-6-(trifluoromethyl)benzylamine
Formula: C₉H₁₀F₃NO
Storage Temperature: Ambient
MDL Number: MFCD09832323
CAS Number: 1017778-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR