2-Methoxy-3-(trifluoromethyl)benzylamine 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC302246-1G PC302246-5G
76613-192EA 118 USD
76613-192 76613-194
2-Methoxy-3-(trifluoromethyl)benzylamine 98%
2-Methoxy-3-(trifluoromethyl)benzylamine
Formula: C₉H₁₀F₃NO
Storage Temperature: Ambient
MDL Number: MFCD09832318
CAS Number: 1017778-87-8
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR