3-Methyl-5-(trifluoromethoxy)benzamide 97%

Supplier: APOLLO SCIENTIFIC

PC302963-1G PC302963-10G PC302963-5G
76599-070EA 390 USD
76599-070 76599-074 76599-072
3-Methyl-5-(trifluoromethoxy)benzamide 97%
3-Methyl-5-(trifluoromethoxy)benzamide
Formula: C₉H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD09025372
CAS Number: 916420-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR