3-Bromo-2-methyl-5-(pentafluorothio)aniline 97%

Supplier: APOLLO SCIENTIFIC

PC303147-1G PC303147-5G
76618-108EA 908 USD
76618-108 76618-106
3-Bromo-2-methyl-5-(pentafluorothio)aniline 97%
3-Bromo-2-methyl-5-(pentafluorothio)aniline
Formula: C₇H₇BrF₅NS
MW: 312.10
Storage Temperature: Ambient
MDL Number: MFCD22201077
CAS Number: 1373920-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR