3-(4-(Difluoromethoxy)phenyl)propionic acid 97%

Supplier: APOLLO SCIENTIFIC

PC302513-1G
76597-972EA 110.02 USD
76597-972
3-(4-(Difluoromethoxy)phenyl)propionic acid 97%
3-(4-(Difluoromethoxy)phenyl)propionic acid
Formula: C₁₀H₁₀F₂O₃
MW: 216.18
Melting Pt: 66…69 °C
Storage Temperature: Ambient
MDL Number: MFCD09863355
CAS Number: 1000932-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR