3-(2-Methoxy-5-(trifluoromethoxy)phenyl)propionic acid 98%

Supplier: APOLLO SCIENTIFIC

PC302502-1G PC302502-5G
76597-934EA 167.06 USD
76597-934 76597-936
3-(2-Methoxy-5-(trifluoromethoxy)phenyl)propionic acid 98%
3-(2-Methoxy-5-(trifluoromethoxy)phenyl)propionic acid
Formula: C₁₁H₁₁F₃O₄
Storage Temperature: Ambient
MDL Number: MFCD09832351
CAS Number: 1017779-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR