5-Fluoro-2-(trifluoromethoxy)benzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

PC303003-1G PC303003-5G PC303003-10G
76599-178EA 70 USD
76599-178 76599-176 76599-180
5-Fluoro-2-(trifluoromethoxy)benzoic acid
5-Fluoro-2-(trifluoromethoxy)benzoic acid
Formula: C₈H₄F₄O₃
MW: 224.11 g/mol
Melting Pt: 89…93 °C
Storage Temperature: Ambient
MDL Number: MFCD11519333
CAS Number: 1092460-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR