5-Methyl-2-(trifluoromethyl)benzonitrile 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC302895-5G PC302895-1G
76626-168EA 142 USD
76626-168 76626-166
5-Methyl-2-(trifluoromethyl)benzonitrile 97%
5-Methyl-2-(trifluoromethyl)benzonitrile
Formula: C₉H₆F₃N
MW: 185.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660290
CAS Number: 886502-61-0
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR