3-(5-Fluoro-2-(trifluoromethoxy)phenyl)propionic acid 98%

Supplier: APOLLO SCIENTIFIC

PC302519-5G PC302519-1G
76597-990EA 508 USD
76597-990 76597-988
3-(5-Fluoro-2-(trifluoromethoxy)phenyl)propionic acid 98%
3-(5-Fluoro-2-(trifluoromethoxy)phenyl)propionic acid
Formula: C₁₀H₈F₄O₃
Storage Temperature: Ambient
MDL Number: MFCD11519341
CAS Number: 1092460-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR