4-Chloro-3-(trifluoromethoxy)benzoic acid 99%

Supplier: APOLLO SCIENTIFIC

Warning

PC302741-1G PC302741-5G PC302741-10G
76598-566EA 232 USD
76598-566 76598-568 76598-570
4-Chloro-3-(trifluoromethoxy)benzoic acid 99%
4-Chloro-3-(trifluoromethoxy)benzoic acid
Formula: C₈H₄ClF₃O₃
MW: 240.57 g/mol
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD06660238
CAS Number: 886500-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR