2-(Dimethylamino)-6-fluorobenzonitrile 97%

Supplier: APOLLO SCIENTIFIC

Warning

PC3009J-5G PC3009J-1G
76625-850EA 210 USD
76625-850 76625-848
2-(Dimethylamino)-6-fluorobenzonitrile 97%
2-(Dimethylamino)-6-fluorobenzonitrile
Formula: C₉H₉FN₂
Boiling Pt: 131…135 °C/5mmHg
Storage Temperature: Ambient
MDL Number: MFCD00052221
CAS Number: 96994-73-9
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR