4'-Fluoro-2'-methylacetophenone

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(4-Fluoro-2-methylphenyl)ethanone

PC302110-1G PC302110-5G PC302110-25G
76596-826EA 14 USD
76596-826 76596-828 76596-830
4'-Fluoro-2'-methylacetophenone
4'-Fluoro-2'-methylacetophenone
Formula: C₉H₉FO
MW: 152.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00028041
CAS Number: 446-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR