3'-Fluoro-2'-methylacetophenone 98%

Supplier: APOLLO SCIENTIFIC

PC302955-10G PC302955-1G PC302955-5G
76599-048EA 309.68 USD
76599-048 76599-052 76599-050
3'-Fluoro-2'-methylacetophenone 98%
3'-Fluoro-2'-methylacetophenone
Formula: C₉H₉FO
MW: 152.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832257
CAS Number: 177942-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR