4'-Chloro-2',6'-difluoroacetophenone 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(4-Chloro-2,6-difluorophenyl)ethanone

PC302972-5G PC302972-1G
76599-100EA 50 USD
76599-100 76599-102
4'-Chloro-2',6'-difluoroacetophenone 98%
4'-Chloro-2',6'-difluoroacetophenone
Formula: C₈H₅ClF₂O
MW: 190.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832259
CAS Number: 1017777-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR