3-Chloro-5-(trifluoromethyl)benzaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC302290-5G PC302290-1G PC302290-25G
76597-366EA 53 USD
76597-366 76597-364 76597-362
3-Chloro-5-(trifluoromethyl)benzaldehyde 97%
3-Chloro-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₄ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD04115845
CAS Number: 477535-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR