α,α,α-Trifluoro-3,5-dinitrotoluene 99%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3,5-Dinitrobenzotrifluoride

PC3018-25G PC3018-100G PC3018-500G
76596-506EA 26 USD
76596-506 76596-508 76596-510
α,α,α-Trifluoro-3,5-dinitrotoluene 99%
α,α,α-Trifluoro-3,5-dinitrotoluene
Formula: C₇H₃F₃N₂O₄
MW: 236.11 g/mol
Boiling Pt: 127…129 °C/9mmHg
Melting Pt: 47…52 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007233
CAS Number: 401-99-0
EINECS: 206-935-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR