2,5-Difluorotoluene 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1,4-Difluoro-2-methylbenzene

PC2940-10G PC2940-25G
76613-084EA 13 USD
76613-084 76613-086
2,5-Difluorotoluene 98%
2,5-Difluorotoluene
Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 115…117 °C
Melting Pt: –35 °C
Density: 1.36 g/cm³ (25 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00000347
CAS Number: 452-67-5
UN: 2388
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR