4-(Difluoromethoxy)benzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC2847O-50G PC2847O-25G PC2847O-5G
76625-824EA 341 USD
76625-824 76625-822 76625-826
4-(Difluoromethoxy)benzonitrile 98%
4-(Difluoromethoxy)benzonitrile
Formula: C₈H₅F₂NO
MW: 169.1282 g/mol
Melting Pt: 40…42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00085006
CAS Number: 90446-25-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR