3-Fluoro-5-methoxybenzyl bromide 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC2797-500MG PC2797-5G PC2797-1G
76622-608EA 60 USD
76622-608 76622-610 76622-612
3-Fluoro-5-methoxybenzyl bromide 97%
3-Fluoro-5-methoxybenzyl bromide
Formula: C₈H₈BrFO
MW: 219.0509 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08741378
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR