4,4'-Dibromooctafluorobiphenyl (DBOB) 99%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4,4-Dibromooctafluorobiphenyl

PC2279-1G PC2279-5G
76617-852EA 55 USD
76617-852 76617-854
4,4'-Dibromooctafluorobiphenyl (DBOB) 99%
4,4'-Dibromooctafluorobiphenyl (DBOB)
Formula: C₁₂Br₂F₈
MW: 455.94 g/mol
Melting Pt: 113…115 °C
Storage Temperature: Ambient
EINECS: 233-847-1
UN: 3152
ADR: 9,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR