4-(Trifluormethyl)nicotinonitrile 95+%

Supplier: APOLLO SCIENTIFIC

Warning

PC2108B-5G PC2108B-10G PC2108B-1G
76625-778EA 118 USD
76625-778 76625-776 76625-774
4-(Trifluormethyl)nicotinonitrile 95+%
4-(Trifluormethyl)nicotinonitrile
Formula: C₇H₃F₃N₂
Boiling Pt: 75…77 °C/15mmHg
Storage Temperature: Ambient
MDL Number: MFCD00153107
CAS Number: 13600-43-6
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR