4-Fluoro-1-naphthonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Fluoronaphthalene-1-carbonitrile

PC2102L-1G
76625-766EA 13 USD
76625-766
4-Fluoro-1-naphthonitrile 97%
4-Fluoro-1-naphthonitrile
Formula: C₁₁H₆FN
MW: 171.18 g/mol
Melting Pt: 89…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00236656
CAS Number: 13916-99-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR