Copper(II) trifluoromethanesulfonate 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Copper triflate
, Copper(II) trifluoromethanesulphonate
, Copper(II) trifluoromethanesulfonate
PC2096E-100G
PC2096E-25G
PC2096E-5G
76622-466EA
18
USD
76622-466
76622-464
76622-468
Copper(II) trifluoromethanesulfonate 98%
Copper (II) trifluoromethanesulphonate
Can produce alkenes at ambient temperatures by catalytic dehydration of alcohols and diols: Helv. Chim. Acta, 70, 607 (1987)
Formula:
C₂CuF₆O₆S₂ MW: 361.69 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077492 CAS Number: 34946-82-2 EINECS: 252-300-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...